2-Chlorisonicotinsäure
Lieferant: Thermo Scientific
Synonyme:
2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure
SPB06649.0010
SPB06649.1
SPB06649EA
MAYBSPB06649.0010EA
0
EUR
MAYBSPB06649.0010
MAYBSPB06649.1
MAYBSPB06649EA
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157.56 g/mol |
MDL:
MFCD00191402 CAS-Nummer: 6313-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...