2-Chlorisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure

M-Clarity™ Qualitätsstufe = MQ100

543918-25G 543918-5G
SIAL543918-25GEA 87.5 EUR
SIAL543918-25G SIAL543918-5G
2-Chlorisonicotinsäure, Sigma-Aldrich®
2-Chlorisonicotinsäure
Formel: C₆H₄ClNO₂
Molecular Weight: 157,56 g/mol
MDL: MFCD00191402
CAS-Nummer: 6313-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR