2-Chlor-3-fluorpyridin

Lieferant: FLUOROCHEM

013882-1G 013882-5G 013882-25G
FLUO013882-1GEA 31.1 EUR
FLUO013882-1G FLUO013882-5G FLUO013882-25G
2-Chlor-3-fluorpyridin
2-Chloro-3-fluoropyridine
Formel: C₅H₃ClFN
MDL: MFCD03095302
CAS-Nummer: 17282-04-1
UN: 2810
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR