2-Chlor-5-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

643556-1G 643556-5G
SIAL643556-1GEA 26 EUR
SIAL643556-1G SIAL643556-5G
2-Chlor-5-fluorpyridin, Sigma-Aldrich®
2-Chloro-5-fluoropyridine
Formel: C₅H₃ClFN
Molecular Weight: 131,54 g/mol
MDL: MFCD03095332
CAS-Nummer: 31301-51-6
UN: 2929
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR