2-Chlor-6-fluorpyridin 98%
Lieferant: Apollo Scientific
PC3083-25G
PC3083-5G
APOSPC3083-25GEA
24.8
EUR
APOSPC3083-25G
APOSPC3083-5G
2-Chlor-6-fluorpyridin 98%
2-Chloro-6-fluoropyridine
Formel:
C₅H₃ClFN |
MDL:
MFCD00077483 CAS-Nummer: 20885-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...