3-Chlor-2,5,6-trifluorpyridin ≥98%

Lieferant: Thermo Scientific
Gefahr

L19543.06 L19543.14
L19543.06EA 73.9 EUR
L19543.06 L19543.14
3-Chlor-2,5,6-trifluorpyridin ≥98%
3-Chloro-2,5,6-trifluoropyridine

Purity: 98+% CAS No.: 2879-42-7 Molecular Formula: C5HClF3N Formula Weight: 167.52 Flash Point: 100°(212°F) Refractive Index: 1.4560 Odour: Unpleasant Boiling Point: 132-134° Beilstein Reference: 1424722 UN No.: 1760 MDL No.: MFCD03001160

Formel: C₅HClF₃N
Siedepunkt: 132…134 °C
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03001160
CAS-Nummer: 2879-42-7
UN: 1760
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR