2-Chlor-4-iodpyridin, Sigma-Aldrich®

Lieferant: Merck

647403-1G 647403-5G
SIAL647403-1GEA 105 EUR
SIAL647403-1G SIAL647403-5G
2-Chlor-4-iodpyridin, Sigma-Aldrich®
2-Chlor-4-iodpyridin
Formel: C₅H₃ClIN
Molecular Weight: 239,44 g/mol
MDL: MFCD01861983
CAS-Nummer: 153034-86-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR