2-Amino-3,5-dichlorpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Dichlorpyridin-2-amin, 3,5-Dichlor-2-pyridylamin

159380050 159380250 159381000
ACRO159380050EA 12.3 EUR
ACRO159380050 ACRO159380250 ACRO159381000
2-Amino-3,5-dichlorpyridin 97%
2-Amino-3,5-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163.01 g/mol
Schmelzpunkt: 80…82 °C
MDL: MFCD00006313
CAS-Nummer: 4214-74-8
EINECS: 224-143-5

Bestellen

Spezifikation Testergebnisse

Appearance Light beige to light brown Powder
Infrared spectrum Conforms
Melting point 79 to 83 °C
Water ≤0.5 %
HPLC ≥96.0 %
Acid insoluble substances ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR