2-Amino-3,5-dichlorpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3,5-Dichlorpyridin-2-amin, 3,5-Dichlor-2-pyridylamin
B21984.14
B21984.22
B21984.14EA
125
EUR
B21984.14
B21984.22
2-Amino-3,5-dichlorpyridin 98%
2-Amino-3,5-dichlorpyridin
Purity: 98%
CAS No.: 4214-74-8
Molecular Formula: C5H4Cl2N2
Formula Weight: 163.01
Odour: Unpleasant
Melting Point: 81-83°
Beilstein Reference: 119376
MDL No.: MFCD00006313
Formel:
C₅H₄Cl₂N₂ Molecular Weight: 163.01 g/mol Schmelzpunkt: 81…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006313 CAS-Nummer: 4214-74-8 EINECS: 224-143-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...