2-Amino-6-chlorpyridin 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chlorpyridin-2-amin, 6-Chlor-2-pyridylamin
L19836.06
L19836.03
L19836.14
L19836.06EA
76.4
EUR
L19836.06
L19836.03
L19836.14
2-Amino-6-chlorpyridin 98%
2-Amino-6-chlorpyridin
Purity: 98%
CAS No.: 45644-21-1
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Melting Point: 69-72°
Beilstein Reference: 108669
Formel:
C₅H₅ClN₂ Molecular Weight: 128,56 g/mol Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234068 CAS-Nummer: 45644-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...