3-Amino-2-chlorpyridin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chlor-3-pyridylamin, 2-Chlorpyridin-3-amin
A15830.09
A15830.18
A15830.30
A15830.09EA
23.6
EUR
A15830.09
A15830.18
A15830.30
3-Amino-2-chlorpyridin ≥98%
3-Amino-2-chlorpyridin
Purity: 98+%
CAS No.: 6298-19-7
Molecular Formula: C5H5ClN2
Formula Weight: 128.56
Flash Point: 185°(365°F)
Odour: Unpleasant
Boiling Point: 130-134°/22mm
Beilstein Reference: 109814
MDL No.: MFCD00006238
Formel:
C₅H₅ClN₂ Molecular Weight: 128,56 g/mol Siedepunkt: 130…134 °C (22 mmHg) Schmelzpunkt: 76…80 °C Flammpunkt: 185 °C (365 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006238 CAS-Nummer: 6298-19-7 EINECS: 228-572-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...