3-Amino-2-chlorpyridin
Lieferant: Thermo Scientific
Synonyme:
2-Chlor-3-pyridylamin, 2-Chlorpyridin-3-amin
SB00803.1
SB00803.10
SB00803.50
MAYBSB00803.1EA
0
EUR
MAYBSB00803.1
MAYBSB00803.10
MAYBSB00803.50
3-Amino-2-chlorpyridin
3-Amino-2-chlorpyridin
Formel:
C₅H₅ClN₂ Molecular Weight: 128,56 g/mol Siedepunkt: 263 °C (1013 hPa) Schmelzpunkt: 78…79 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006238 CAS-Nummer: 6298-19-7 EINECS: 228-572-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...