4-Amino-3,5-dichlorpyridin ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3,5-Dichlor-4-pyridylamin, 3,5-Dichlorpyridin-4-amin

B24725.06 B24725.14
B24725.06EA 31.4 EUR
B24725.06 B24725.14
4-Amino-3,5-dichlorpyridin ≥98%
4-Amino-3,5-dichlorpyridin

Purity: 98+% CAS No.: 22889-78-7 Molecular Formula: C5H4Cl2N2 Formula Weight: 163.01 Melting Point: 159-163° Beilstein Reference: 120547

Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00125023
CAS-Nummer: 22889-78-7
EINECS: 245-304-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR