4-Amino-3,5-dichlorpyridin

Lieferant: Thermo Scientific

Synonyme: 3,5-Dichlor-4-pyridylamin, 3,5-Dichlorpyridin-4-amin

RF01230.1 RF01230.10
MAYBRF01230.1EA 0 EUR
MAYBRF01230.1 MAYBRF01230.10
4-Amino-3,5-dichlorpyridin
4-Amino-3,5-dichlorpyridin
Formel: C₅H₄Cl₂N₂
Molecular Weight: 163,01 g/mol
MDL: MFCD00125023
CAS-Nummer: 22889-78-7
EINECS: 245-304-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR