2-Chlor-3-hydroxypyridin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Chlor-3-pyridinol
A11376.14
A11376.22
A11376.14EA
127
EUR
A11376.14
A11376.22
2-Chlor-3-hydroxypyridin ≥98%
2-Chlor-3-hydroxypyridin
Purity: 98%
CAS No.: 6636-78-8
Molecular Formula: C5H4ClNO
Formula Weight: 129.55
Melting Point: 168-172°
Beilstein Reference: 2216
Formel:
C₅H₄ClNO Molecular Weight: 129,55 g/mol Siedepunkt: 212 °C (1013 hPa) Schmelzpunkt: 168…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006235 CAS-Nummer: 6636-78-8 EINECS: 229-635-3 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...