6-Chlor-2-hydroxypyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Chlor-6-pyridinol, 6-Chlor-2-pyridinol, 2-Chlor-6-hydroxypyridin

A13164.14 A13164.22
A13164.14EA 60 EUR
A13164.14 A13164.22
6-Chlor-2-hydroxypyridin ≥98%
6-Chlor-2-hydroxypyridin

Purity: 98% CAS No.: 16879-02-0 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Flash Point: 163°(325°F) Melting Point: 129-131° Beilstein Reference: 108665 MDL No.: MFCD00006278

Formel: C₅H₄ClNO
Molecular Weight: 129.55 g/mol
Schmelzpunkt: 129…131 °C
Flammpunkt: 163 °C (325 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006278
CAS-Nummer: 16879-02-0
EINECS: 240-909-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR