5-Chlor-3-hydroxypyridin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L07568.06 L07568.14
L07568.06EA 43 EUR
L07568.06 L07568.14
5-Chlor-3-hydroxypyridin ≥99%
5-Chlor-3-hydroxypyridin

Purity: 99% CAS No.: 74115-12-1 Molecular Formula: C5H4ClNO Formula Weight: 129.55 Melting Point: 159-163° Beilstein Reference: 1422550

Formel: C₅H₄ClNO
Schmelzpunkt: 159…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006379
CAS-Nummer: 74115-12-1
EINECS: 277-727-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR