5-Chlor-3-hydroxypyridin
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Chlor-3-pyridinol, 3-Chlor-5-hydroxypyridin, 3-Chlor-5-pyridinol
SB00891.1
SB00891EA
SB00891EE
MAYBSB00891.1EA
0
EUR
MAYBSB00891.1
MAYBSB00891EA
MAYBSB00891EE
5-Chlor-3-hydroxypyridin
5-Chlor-3-hydroxypyridin
Formel:
C₅H₄ClNO Molecular Weight: 129.55 g/mol Schmelzpunkt: 158 |
MDL:
MFCD00006379 CAS-Nummer: 74115-12-1 EINECS: 277-727-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...