2-Pyridincarbonitril ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Pyridincarbonsäurenitril, 2-Cyanpyridin
A12900.22
A12900.36
A12900.22EA
51.4
EUR
A12900.22
A12900.36
2-Pyridincarbonitril ≥99%
2-Pyridincarbonitril
Purity: 99%
CAS No.: 100-70-9
Molecular Formula: C6H4N2
Formula Weight: 104.11
Flash Point: 89°(192°F)
Refractive Index: 1.5290
Odour: Characteristic
pH: 8.4
Melting Point: 26-28°
Boiling Point: 212-214°
Beilstein Reference: 107710
MDL No.: MFCD00006218
Formel:
C₆H₄N₂ Molecular Weight: 104.11 g/mol Siedepunkt: 212…214 °C Schmelzpunkt: 26…28 °C Dichte: 1.081 Flammpunkt: 89 °C (192 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006218 CAS-Nummer: 100-70-9 EINECS: 202-880-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...