Isonicotinonitril ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Cyanpyridin, 4-Pyridincarbonitril, 4-Pyridincarbonsäurenitril

A10286.09 A10286.0E A10286.22 A10286.36
A10286.09EA 27.4 EUR
A10286.09 A10286.0E A10286.22 A10286.36
Isonicotinonitril ≥98%
Isonicotinonitril

Purity: 98% CAS No.: 100-48-1 Molecular Formula: C6H4N2 Formula Weight: 104.11 Flash Point: 88°(190°F) Odour: Pungent pH: 6 Melting Point: 76-80° Boiling Point: 194-196° Beilstein Reference: 107712 MDL No.: MFCD00006417

Formel: C₆H₄N₂
Molecular Weight: 104,11 g/mol
Siedepunkt: 194…196 °C
Schmelzpunkt: 76…80 °C
Flammpunkt: 88 °C (190 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006417
CAS-Nummer: 100-48-1
EINECS: 202-856-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR