Ethylisonicotinat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Picolinic acid ethyl ester, 4-Pyridinecarboxylic acid ethyl ester, Pyridine-4-carboxylic acid ethyl ester, Ethyl 4-pyridinecarboxylate, Isonicotinic acid ethyl ester

L03010.22 L03010.36
L03010.22EA 46.1 EUR
L03010.22 L03010.36
Ethylisonicotinat ≥98%
Ethylisonicotinat

Purity: 98% CAS No.: 1570-45-2 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 87°(188°F) Refractive Index: 1.5010 Odour: Characteristic Boiling Point: 220° Beilstein Reference: 122942 Merck Reference: 14,5187 MDL No.: MFCD00006428

Formel: C₈H₉NO₂
Molecular Weight: 151.16 g/mol
Siedepunkt: 220 °C
Dichte: 1.008
Flammpunkt: 87 °C (188 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006428
CAS-Nummer: 1570-45-2
EINECS: 216-379-2
Merck Index: 13,05206

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR