2-Aminonicotinsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Aminopyridin-3-carbonsäure, 2-Amino-3-pyridincarbonsäure
A11195.14
A11195.06
A11195.22
A11195.14EA
120
EUR
A11195.14
A11195.06
A11195.22
2-Aminonicotinsäure ≥98%
2-Aminonicotinsäure
Purity: 98+%
CAS No.: 5345-47-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: ca 298° dec.
Beilstein Reference: 119031
Formel:
C₆H₆N₂O₂ Molecular Weight: 138.13 g/mol Schmelzpunkt: ∼ 298 °C (Zers.) Dichte: 1.545 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006318 CAS-Nummer: 5345-47-1 EINECS: 226-296-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...