6-Hydroxynicotinsäure
Lieferant: Thermo Scientific
Synonyme:
6-Hydroxypyridin-3-carbonsäure, 6-Hydroxy-3-pyridincarbonsäure
SB00737.1
SB00737.10
SB00737.50
MAYBSB00737.1EA
0
EUR
MAYBSB00737.1
MAYBSB00737.10
MAYBSB00737.50
6-Hydroxynicotinsäure
6-Hydroxynicotinsäure
Formel:
C₆H₅NO₃ Molecular Weight: 139,11 g/mol Schmelzpunkt: 304…306 °C |
MDL:
MFCD00006277 CAS-Nummer: 5006-66-6 EINECS: 225-682-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...