6-Hydroxynicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 6-Hydroxypyridin-3-carbonsäure, 6-Hydroxy-3-pyridincarbonsäure

009265-25G 009265-100G
FLUO009265-25GEA 34.1 EUR
FLUO009265-25G FLUO009265-100G
6-Hydroxynicotinsäure
6-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Schmelzpunkt: 304…306 °C
MDL: MFCD00006277
CAS-Nummer: 5006-66-6
EINECS: 225-682-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR