2,3,6-Trifluorpyridin ≥97%

Lieferant: Thermo Scientific
Achtung

L19540.03 L19540.06
L19540.03EA 69.5 EUR
L19540.03 L19540.06
2,3,6-Trifluorpyridin ≥97%
2,3,6-Trifluoropyridine

Purity: 97% CAS No.: 3512-18-3 Molecular Formula: C5H2F3N Formula Weight: 133.07 Flash Point: 30°(86°F) Refractive Index: 1.4200 Boiling Point: 100-102° Beilstein Reference: 1365146 UN No.: 1993 MDL No.: MFCD03001158

Formel: C₅H₂F₃N
Molecular Weight: 133,07 g/mol
Siedepunkt: 100…102 °C
Dichte: 1,402
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD03001158
CAS-Nummer: 3512-18-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR