2-Fluornicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluorpyridin-3-carbonsäure, 2-Fluor-3-pyridincarbonsäure

005467-1G 005467-5G 005467-25G
FLUO005467-1GEA 36.9 EUR
FLUO005467-1G FLUO005467-5G FLUO005467-25G
2-Fluornicotinsäure
2-Fluornicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD00040744
CAS-Nummer: 393-55-5
EINECS: 206-888-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR