2-Amino-5-fluorpyridin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
L20010.03
L20010.06
L20010.03EA
79.6
EUR
L20010.03
L20010.06
2-Amino-5-fluorpyridin 97%
2-Amino-5-fluorpyridin
Purity: 97%
CAS No.: 21717-96-4
Molecular Formula: C5H5FN2
Formula Weight: 112.11
Odour: Characteristic
Melting Point: 93-97°
Beilstein Reference: 385938
UN No.: 2811
MDL No.: MFCD01861120
Formel:
C₅H₅FN₂ Molecular Weight: 112,11 g/mol Schmelzpunkt: 93…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861120 CAS-Nummer: 21717-96-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...