2-Amino-6-fluorpyridin

Lieferant: FLUOROCHEM
Achtung

024171-1G 024171-5G
FLUO024171-1GEA 30.8 EUR
FLUO024171-1G FLUO024171-5G
2-Amino-6-fluorpyridin
2-Amino-6-fluorpyridin
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD04114200
CAS-Nummer: 1597-32-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR