4-(Dimethylamino)pyridin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
DMAP, N,N-Dimethylpyridin-4-amin
A13016.06
A13016.14
A13016.22
A13016.06EA
21.8
EUR
A13016.06
A13016.14
A13016.22
4-(Dimethylamino)pyridin 99%
4-(Dimethylamino)pyridin
Purity: 99%
CAS No.: 1122-58-3
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 124°(255°F)
Physical Form: Crystals or powder
Melting Point: 110-113°
Boiling Point: 162°/50mm
Beilstein Reference: 110354
UN No.: 2928
MDL No.: MFCD00006418
Formel:
C₇H₁₀N₂ Siedepunkt: 162 °C (50 mmHg) Schmelzpunkt: 110…113 °C Flammpunkt: 124 °C (255 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006418 CAS-Nummer: 1122-58-3 EINECS: 214-353-5 UN: 2928 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...