2-Amino-5-nitropyridin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine
A11352.14
A11352.22
A11352.14EA
63.3
EUR
A11352.14
A11352.22
2-Amino-5-nitropyridin ≥97%
2-Amino-5-nitropyridin
Purity: 97%
CAS No.: 4214-76-0
Molecular Formula: C5H5N3O2
Formula Weight: 139.11
Flash Point: 224°(435°F)
Melting Point: 185-190°
Beilstein Reference: 120353
MDL No.: MFCD00006325
Formel:
C₅H₅N₃O₂ Molecular Weight: 139,11 g/mol Schmelzpunkt: 185…190 °C Dichte: 1,623 g/cm³ (20 °C) Flammpunkt: 224 °C (435 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006325 CAS-Nummer: 4214-76-0 EINECS: 224-145-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...