2-Amino-5-nitropyridin
Lieferant: FLUOROCHEM
|
Achtung
|
Synonyme:
5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine
010697-25G
010697-100G
FLUO010697-25GEA
42.9
EUR
FLUO010697-25G
FLUO010697-100G
2-Amino-5-nitropyridin
2-Amino-5-nitropyridin
Formel:
C₅H₅N₃O₂ Molecular Weight: 139.11 g/mol Schmelzpunkt: 185…188 °C Dichte: 1.623 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006325 CAS-Nummer: 4214-76-0 EINECS: 224-145-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...