4-Amino-3-nitropyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine
M-Clarity™ Qualitätsstufe = MQ100
646962-10G
646962-1G
SIAL646962-10GEA
197
EUR
SIAL646962-10G
SIAL646962-1G
4-Amino-3-nitropyridin, Sigma-Aldrich®
4-Amino-3-nitropyridin
Formel:
C₅H₅N₃O₂ Molecular Weight: 139,11 g/mol |
MDL:
MFCD00160304 CAS-Nummer: 1681-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...