2-Amino-3-hydroxypyridin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Amino-3-pyridinol

153570250 153571000
ACRO153570250EA 54.3 EUR
ACRO153570250 ACRO153571000
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Siedepunkt: 250 °C (1013 hPa)
Schmelzpunkt: 170…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006317
CAS-Nummer: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Beige to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 170 to 176 °C
Water ≤0.5 % (K.F.)
Additional info residue on sublimation: 2%
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 % (On anhydrous substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR