4-Methoxypyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

460621-25ML 460621-5ML
SIAL460621-25MLEA 298 EUR
SIAL460621-25ML SIAL460621-5ML
4-Methoxypyridin, Sigma-Aldrich®
4-Methoxypyridine
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00674049
CAS-Nummer: 620-08-6
EINECS: 210-624-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR