2-Hydroxypyridin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Pyridinol
A14522.14
A14522.22
A14522.36
A14522.14EA
31.7
EUR
A14522.14
A14522.22
A14522.36
2-Hydroxypyridin ≥98%
2-Hydroxypyridin
Purity: 98%
CAS No.: 142-08-5
Molecular Formula: C5H5NO
Formula Weight: 95.10
Storage Temperature: 2°C to 8°C
Flash Point: 210°(410°F)
Odour: Recognizable
Melting Point: 102-111°
Boiling Point: 280-281°
Beilstein Reference: 105786
UN No.: 2811
MDL No.: MFCD00006268
Formel:
C₅H₅NO Molecular Weight: 95,10 g/mol Siedepunkt: 280…281 °C Schmelzpunkt: 102…111 °C Dichte: 1,39 Flammpunkt: 210 °C (410 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00006268 CAS-Nummer: 142-08-5 EINECS: 205-520-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...