2-Brom-3-methylpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Bromo-3-picoline

448109-5ML 448109-25ML
SIAL448109-5MLEA 91.1 EUR
SIAL448109-5ML SIAL448109-25ML
2-Brom-3-methylpyridin, Sigma-Aldrich®
2-Brom-3-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172,02 g/mol
MDL: MFCD00239380
CAS-Nummer: 3430-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR