4-Brom-2-methylpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

662739-1G 662739-250MG
SIAL662739-1GEA 105 EUR
SIAL662739-1G SIAL662739-250MG
4-Brom-2-methylpyridin, Sigma-Aldrich®
4-Brom-2-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172,02 g/mol
MDL: MFCD03788196
CAS-Nummer: 22282-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR