2-Amino-3-brom-5-methylpyridin

Lieferant: FLUOROCHEM
Achtung

CFF291.25 002493-1G 002493-5G 002493-25G
CHMPCFF291.25EA 532 EUR
CHMPCFF291.25 FLUO002493-1G FLUO002493-5G FLUO002493-25G
2-Amino-3-brom-5-methylpyridin
2-Amino-3-brom-5-methylpyridin
Formel: C₆H₇BrN₂
MDL: MFCD00068231
CAS-Nummer: 17282-00-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR