2-Amino-5-brom-3-methylpyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Amino-5-brom-3-picolin, 5-Brom-3-methyl-2-pyridylamin, 5-Brom-3-methylpyridin-2-amin

002494-1G 002494-5G
FLUO002494-1GEA 31.1 EUR
FLUO002494-1G FLUO002494-5G
2-Amino-5-brom-3-methylpyridin
2-Amino-5-brom-3-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187.04 g/mol
MDL: MFCD00068232
CAS-Nummer: 3430-21-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR