2-Amino-5-brom-3-methylpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 2-Amino-5-brom-3-picolin, 5-Brom-3-methyl-2-pyridylamin, 5-Brom-3-methylpyridin-2-amin

M-Clarity™ Qualitätsstufe = MQ100

525537-1G 525537-5G
SIAL525537-1GEA 33.1 EUR
SIAL525537-1G SIAL525537-5G
2-Amino-5-brom-3-methylpyridin, Sigma-Aldrich®
2-Amino-5-brom-3-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187,04 g/mol
MDL: MFCD00068232
CAS-Nummer: 3430-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR