2-Amino-5-brom-4-methylpyridin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline
M-Clarity™ Qualitätsstufe = MQ100
576786-1G
576786-25G
576786-5G
SIAL576786-1GEA
31.7
EUR
SIAL576786-1G
SIAL576786-25G
SIAL576786-5G
2-Amino-5-brom-4-methylpyridin, Sigma-Aldrich®
2-Amino-5-brom-4-methylpyridin
Formel:
C₆H₇BrN₂ Molecular Weight: 187,04 g/mol |
MDL:
MFCD03427660 CAS-Nummer: 98198-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...