6-Amino-3-brom-2-methylpyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Brom-6-methylpyridin-2-amin, 2-Amino-5-brom-6-methylpyridin, 3-Brom-2-methyl-6-pyridylamin, 3-Brom-2-methylpyridin-6-amin, 5-Brom-6-methyl-2-pyridylamin, 6-Amino-3-brom-2-picolin, 2-Amino-5-brom-6-picolin
A12948.06
A12948.14
A12948.06EA
45.3
EUR
A12948.06
A12948.14
6-Amino-3-brom-2-methylpyridin 97%
6-Amino-3-brom-2-methylpyridin
Purity: 97%
CAS No.: 42753-71-9
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Melting Point: 80-82°
Beilstein Reference: 114140
Formel:
C₆H₇BrN₂ Molecular Weight: 187,04 g/mol Schmelzpunkt: 80…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00068230 CAS-Nummer: 42753-71-9 EINECS: 255-927-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...