6-Amino-3-brom-2-methylpyridin

Lieferant: FLUOROCHEM
Achtung

Synonyme: 5-Brom-6-methylpyridin-2-amin, 2-Amino-5-brom-6-methylpyridin, 3-Brom-2-methyl-6-pyridylamin, 3-Brom-2-methylpyridin-6-amin, 5-Brom-6-methyl-2-pyridylamin, 6-Amino-3-brom-2-picolin, 2-Amino-5-brom-6-picolin

002495-1G 002495-5G 002495-25G 002495.25
FLUO002495-1GEA 31.1 EUR
FLUO002495-1G FLUO002495-5G FLUO002495-25G FLUO002495.25
6-Amino-3-brom-2-methylpyridin
6-Amino-3-brom-2-methylpyridin
Formel: C₆H₇BrN₂
Molecular Weight: 187.04 g/mol
MDL: MFCD00068230
CAS-Nummer: 42753-71-9
EINECS: 255-927-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR