2-Acetylpyridin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Pyridylmethylketon, Methyl-2-pyridylketon

A12688.14 A12688.22 A12688.36
A12688.14EA 41.9 EUR
A12688.14 A12688.22 A12688.36
2-Acetylpyridin ≥98%
2-Acetylpyridin

Purity: 98% CAS No.: 1122-62-9 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 76°(169°F) Refractive Index: 1.5220 Odour: Unpleasant Melting Point: 8-10° Boiling Point: 189-190° Beilstein Reference: 107759 MDL No.: MFCD00006303

Formel: C₇H₇NO
Molecular Weight: 121.14 g/mol
Siedepunkt: 189…190 °C
Schmelzpunkt: 8…10 °C
Dichte: 1.082
Flammpunkt: 76 °C (169 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006303
CAS-Nummer: 1122-62-9
EINECS: 214-355-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR