2-Fluor-3-methylpyridin

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2-Fluor-3-picolin

001764-1G 001764-25G
FLUO001764-1GEA 32 EUR
FLUO001764-1G FLUO001764-25G
2-Fluor-3-methylpyridin
2-Fluor-3-methylpyridin
Formel: C₆H₆FN
Molecular Weight: 111.12 g/mol
MDL: MFCD00041223
CAS-Nummer: 2369-18-8
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR