2-Fluor-4-methylpyridin ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Fluor-4-picolin
L01602.06
L01602.03
L01602.06EA
87.8
EUR
L01602.06
L01602.03
2-Fluor-4-methylpyridin ≥98%
2-Fluor-4-methylpyridin
Purity: 98%
CAS No.: 461-87-0
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 51°(123°F)
Refractive Index: 1.4725
Boiling Point: 155-156°
Beilstein Reference: 107086
UN No.: 1993
MDL No.: MFCD00041224
Formel:
C₆H₆FN Molecular Weight: 111,12 g/mol Siedepunkt: 155…156 °C Dichte: 1,078 Flammpunkt: 51 °C (123 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041224 CAS-Nummer: 461-87-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...