2-Fluor-6-methylpyridin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Fluor-6-picolin
L01561.06
L01561.03
L01561.06EA
58
EUR
L01561.06
L01561.03
2-Fluor-6-methylpyridin ≥98%
2-Fluor-6-methylpyridin
Purity: 98%
CAS No.: 407-22-7
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 66°(150°F)
Refractive Index: 1.4700
Boiling Point: 140-141°
Beilstein Reference: 107084
MDL No.: MFCD00041226
Formel:
C₆H₆FN Molecular Weight: 111.12 g/mol Siedepunkt: 140…141 °C Dichte: 1.077 Flammpunkt: 66 °C (150 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041226 CAS-Nummer: 407-22-7 EINECS: 206-980-8 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...