2-Fluor-6-methylpyridin

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2-Fluor-6-picolin

006288-5G 006288-25G 006288-100G
FLUO006288-5GEA 0 EUR
FLUO006288-5G FLUO006288-25G FLUO006288-100G
2-Fluor-6-methylpyridin
2-Fluor-6-methylpyridin
Formel: C₆H₆FN
Molecular Weight: 111,12 g/mol
MDL: MFCD00041226
CAS-Nummer: 407-22-7
EINECS: 206-980-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR