2-(2-Aminoethyl)pyridin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(2-Pyridyl)ethylamin

L05627.03 L05627.06 L05627.14
L05627.03EA 30.8 EUR
L05627.03 L05627.06 L05627.14
2-(2-Aminoethyl)pyridin 98%
2-(2-Aminoethyl)pyridin

Purity: 98% CAS No.: 2706-56-1 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 100°(212°F) Refractive Index: 1.5360 Odour: Amine-like Boiling Point: 208° Beilstein Reference: 111208 UN No.: 2735 MDL No.: MFCD00006367

Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
Siedepunkt: 208 °C
Dichte: 1,020
Flammpunkt: 100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006367
CAS-Nummer: 2706-56-1
EINECS: 220-295-1
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR