2-Amino-5-methylpyridin
Lieferant: Thermo Scientific
Synonyme:
5-Methylpyridin-2-amin, 2-Amino-5-picolin, 5-Methyl-2-pyridylamin
SB01317.1
SB01317EA
SB01317.50
MAYBSB01317.1EA
0
EUR
MAYBSB01317.1
MAYBSB01317EA
MAYBSB01317.50
2-Amino-5-methylpyridin
2-Amino-5-methylpyridin
Formel:
C₆H₈N₂ Molecular Weight: 108,14 g/mol Siedepunkt: 227,1 °C (1013 hPa) Schmelzpunkt: 74…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006328 CAS-Nummer: 1603-41-4 EINECS: 216-503-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...